Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brucella Medium Base (Dehydrated)

Catalog No. OXCM0169B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0169B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0169B Supplier Thermo Scientific™ Supplier No. CM0169B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and cultivate Brucella with this medium when used with Brucella Selective Supplement (SR0083 or SR0209).

Isolate and cultivate Brucella using Thermo Scientific™ Oxoid™ Brucella Medium Base (Dehydrated) when used with Brucella Selective Supplement (SR0083 or SR0209). Brucella medium Base may be used to prepare the serum-dextrose-antibiotic medium as described by Jones and Brinley Morgan1 for the cultivation and isolation of Brucella, including fastidious types.

Isolate and cultivate Brucella using Brucella Medium Base when used with Brucella Selective Supplement (SR0083 or SR0209).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Brucella Medium Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.1L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.