Learn More
Brucine-Sulfanilic Acid Solution, for Nitrate Analysis, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Brucine-Sulfanilic Acid Solution |
| Color | Colorless to Pink |
| CAS | 7732-18-5, 7647-01-0, 5787-00-8, 121-57-3, 7732-18-5 |
| CAS Min % | 94.0, 3.32, 0.94, 0.09 |
| CAS Max % | 97.0, 3.45, 0.98, 0.1 |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| pH | <1 |
| Quantity | 500 mL |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.