Learn More
Microbiologics™ Bruker MALDI Biotyper System Extended Identification Panel (IQ/OQ)
Shop All Microbiologics Products

Description
Includes
Bacillus subtilis derived from ATCC™ 6051™†, Brevundimonas vesicularis derived from ATCC™ 11426™, Chryseobacterium indologenes derived from ATCC™ 29897™, Curtobacterium pusillum derived from ATCC™ 19096™, Deinococcus radiophilus derived from ATCC™ 27603™, Enterococcus faecalis derived from ATCC™ 19433™, Methylobacterium organophilum derived from ATCC™ 27886™, Micrococcus luteus derived from ATCC™ 4698™, Proteus vulgaris derived from ATCC™ 29905™, Pseudomonas aeruginosa derived from ATCC™ 10145™, Ralstonia pickettii derived from ATCC™ 27511™, Sphingomonas paucimobilis derived from ATCC™ 29837™, Staphylococcus aureus derived from ATCC™ 12600™, Staphylococcus epidermidis derived from ATCC™ 14990™, Stenotrophomonas maltophilia derived from ATCC™ 13637™
Specifications
Specifications
| ATCC® | ATCC™ 6051™, ATCC™ 11426™, ATCC™ 29897™, ATCC™ 19096™, ATCC™ 27603™, ATCC™ 19433™, ATCC™ 4698™, ATCC™ 29905™, ATCC™ 10145™, ATCC™ 27511™, ATCC™ 29837™, ATCC™ 12600™, ATCC™ 14990™, ATCC™ 13637™ |
| Biosafety Level | 2 |
| Description | Bruker MALDI Biotyper System Extended Identification Panel (IQ/OQ) |
| Detects | Bacillus subtilis derived from ATCC™ 6051™, Brevundimonas vesicularis derived from ATCC™ 11426™, Chryseobacterium indologenes derived from ATCC™ 29897™, Curtobacterium pusillum derived from ATCC™ 19096™, Deinococcus radiophilus derived from ATCC™ 27603™, Enterococcus faecalis derived from ATCC™ 19433™, Methylobacterium organophilum derived from ATCC™ 27886™, Micrococcus luteus derived from ATCC™ 4698™, Proteus vulgaris derived from ATCC™ 29905™, Pseudomonas aeruginosa derived from ATCC™ 10145™, Ralstonia pickettii derived from ATCC™ 27511™, Sphingomonas paucimobilis derived from ATCC™ 29837™, Staphylococcus aureus derived from ATCC™ 12600™, Staphylococcus epidermidis derived from ATCC™ 14990™, Stenotrophomonas maltophilia derived from ATCC™ 13637™ |
| Format | KWIK-STIK™ Two Pack |
| For Use With (Application) | Molecular Testing |
| Quantity | 2 Pk. |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.