Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Bruker MALDI Biotyper System Food Pathogens Identification Panel
SDP

Catalog No. 23045548 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
8 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-045-548 8 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-045-548 Supplier Microbiologics™ Supplier No. 5255P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiologics offers a wide range of bundled QC Sets and Panels for the most commonly used technologies and standards. Each set or panel contains the manufacturer-recommended quality control strains for a particular instrument or test method in a simple, ready-to-use format.

Bruker MALDI Biotyper System Food Pathogens Identification Panel contains two each of Campylobacter jejuni subsp. jejuni derived from ATCC™ 33291™,Cronobacter sakazakii derived from ATCC™ 29544™,Listeria monocytogenes (4b) derived from ATCC™ 13932™,and Salmonella enterica subsp. enterica serovar Typhimurium derived from ATCC™ 14028™

Specifications

Content And Storage 2°C to 8°C
Format QC Sets and Panels: KWIK-STIK™
Product Type Quality Control
For Use With (Application) Microbial Identification
No. per Pack 2
Packaging Type Plastic Canister
Quantity 8 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.