Learn More
Sigma Aldrich Fine Chemicals Biosciences ZapCapR bottle-top filters
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2226071PAK
Encompass
Whatman(R) ZapCap(R) CR Bottle Top filtersZapCap(R)for filtration of medium volumes cell culture media and HPLC SOLUTIONons.features and benefitsComplete 500 ml units with tubing nozzle for attaching to bottles (bottle-top)Connection seals on any standard bottles 33 to 45 mmMembrane diameter 76 mm filter area 39.2 cm2ZapCap-S with included borosilicate prefilter for high flow ratesZapCap-S Plus with integral borosilicate prefilter for very high flow ratesZapCap-CR the chemical-resistant bottle-top filterCan be used up to 50AºCZapCap-CRfiltration of HPLC SOLUTIONons1. Polyamide membrane filters (NYL) for filtering down to 0.2 mum in HPLC/fPLC SOLUTIONons when the column packing is 10 mum 2. PTfE membrane filters for filtering down to 0.45mum in organic SOLUTIONons strong acids or aldehydes ZapCap-Sfiltration of cell culture media1. Cellulose acetate membrane filters (CA) with extremely low protein binding for cell culture media and other aqueous SOLUTIONons2.
Catalog No. 50-290-0396
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.