Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRUNOL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561393 Supplier Invitrogen™ Supplier No. PA561393
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLPGLPAPIG VNGFGPLTPQ TNGQPGSDTL YNNGLS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

RNA-binding protein implicated in the regulation of pre-mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of TNNT2 in a muscle-specific splicing enhancer (MSE)-dependent manner. Promotes also exon exclusion of INSR pre-mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRUNOL6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CELF6
Gene Accession No. Q96J87
Gene Alias 6330569O16Rik; Bruno -like 6, RNA binding protein; BRUNOL6; bruno-like 6, RNA binding protein; bruno-like protein 6; CELF6; CELF-6; CUG-BP and ETR-3 like factor 6; CUG-BP- and ETR-3-like factor 6; CUGBP Elav-like family member 6; CUGBP, Elav-like family member 6; RNA-binding protein BRUNOL-6
Gene Symbols CELF6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRUNOL6. Recombinant protein control fragment (Product #RP-100787).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60677
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.