Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ FlexBrush™ and Covers
SDP

Catalog No. 19165650 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-165-650 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-165-650 Supplier Micronova™ Supplier No. FB12
  • $986.00
    $697.65 / Case of 12

    Save $288.35

    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These S and J-shaped nylon bristle brushes feature a flexible core that allows the user to shape the brush for custom applications, making them ideal for cleaning pipe exteriors, fittings and railings.

  • Acme threaded end
  • Not autoclavable
  • Polyester Bristles
  • Twisted Zinc
  • Plastic Handle

Specifications

Product Type Brush
Bristle Type Medium/Stiff
Color-coded No
Handle Material Plastic
Handle Type Acme Thread
Tip Type Bent
Sterility Non-sterile
Autoclavable Not Autoclavable
Color White
Disposable Reusable
For Use With (Application) Easily cleans overhead pipes, coving and deep
Length (English) Brush 12 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.