Learn More
PIP™ Brush Fire Helmet with ESS Goggle Mounts
Catalog No. 19046416 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Red
White
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-046-416 Yellow
19-046-414 Red
19-046-415 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-046-416 Supplier PIP™ Supplier No. 8696081
PIP™ Brush Fire Helmet with ESS Goggle Mounts features a lightweight Aramid fibers composite resin shell for excellent strength and durability
  • Composite resins offer higher temperature resistance (up to 500°F) without distorting or melting
  • Shell weighs less than Thermoplastic equivalent models
  • Polyurethane impact liner with leather sweatband provides impact and thermal protection
  • Headband with removable padding for cleaning or replacement
  • Ratcheting adjustable headband with merino padding for fit and comfort
  • Leather reinforced mesh 6-point ribbon cradle
  • ESS goggle mounts
  • 3-Point Nomex™ chin strap with quick-release buckle keeps helmet secure
  • High-gloss paint finish available in a variety of colors
  • Meets ANSI/ISEA Z89.1-2009 requirements
  • Certified to NFPA 1971, AS 4067
Specifications
Color Yellow
Product Type Fire Protectant Helmets
Size One Size
Warning - Prop 65 - Cancer and Reproductive Harm