Learn More
Enterprise Technology Solutions HK Surgical Monty Infiltration Cannula Set (#IC-MSET)
SDP
Supplier:  Enterprise Technology Solutions HKSICMSET14
Encompass_Preferred
he HK Surgical Monty Infiltration Cannula Set (#IC-MSET) includes five cannulas total: two Full Monty Cannulas, two Half Monty Cannulas, and a Tip Monty Cannula.Monty Infiltrators are innovative tumescent infiltration cannulas that dramatically improve the technique for infiltration of tumescent local anesthesia. Using a Monty Infiltrator almost entirely eliminates the discomfort patients experienced with other infiltration techniques. The routine IM narcotic analgesia for tumescent infiltration is a thing of the past.Monty Infiltrators are 15 cm or 20 cm long, blunt-tipped cannulas, available in 18, 16, or 14 Gauge diameters (or tumescent liposuction entirely by local anesthesia, 16 Gauge, 15 cm Monty Infiltrators are preferred).
Catalog No. 50-240-7142
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.