Learn More
Andwin Scientific BSA 30% 50ML
SDP
Supplier:  Andwin Scientific 126625
Encompass
The Albumin, Bovine Serum is a 30% aqueous solution made to be used for vitro assays. It is a liquid chemical that is classified as nonhazardous for it does not contain any hazardous ingredients or substances. This product is packaged and sold in a 50 mL glass bottle.
Catalog No. NC9387846
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.