Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals BSA Blocking Buffer, 3% in PBS

Catalog No. p-7082281
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61655AK 250 mL
AAJ61655AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61655AK Supplier Thermo Scientific Chemicals Supplier No. J61655AK
Only null left
Add to Cart

BSA (bovine serum albumin) blocking buffer, 3% in PBS, is a blocking agent ideal for saturating excess protein-binding sites on membranes.

BSA blocking buffer have a high affinity for proteins. A variety of blocking buffers ranging from milk or normal serum to highly purified proteins have been used to block free sites on a membrane. It is a good blocking agent that is easily prepared in the lab. It can be used in a range from 0.3% to 5%. It is also useful for saturating excess protein-binding sites on the membranes and microplates in immunoassays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• BSA blocking buffer contains a serum albumin protein isolated from a cow with a high affinity for proteins

Application

• BSA (bovine serum albumin) blocking buffer is used as a blocking reagent ideal for saturating excess protein-binding sites on membranes and microplates for Western blotting and ELISA applications
• This buffer can be used in a range from 0.3% to 5% dissolved in PBS solution

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
Concentration 3% in PBS
Physical Form Liquid
Quantity 250 mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.