Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals BSA Blocking Buffer, 5% in TBS, with 0.05% Tween-20

Catalog No. AAJ60098AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60098AP 500 mL
AAJ60098AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60098AP Supplier Thermo Scientific Chemicals Supplier No. J60098AP
Add to Cart

missing translation for 'validMainframeMsg'

BSA blocking buffer, 5% in TBS, with 0.05% Tween-20 is used for saturating excess protein-binding sites on the membranes and microplates in immunoassays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
Physical Form Liquid
Quantity 500 mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.