Learn More
TARGETMOL CHEMICALS INC BSH-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T10623
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BSH-IN-1 is a potent and covalent inhibitor of gut bacterial recombinant bile salt hydrolases (BSHs; IC50s 108 nM and 427 nM for B. longum BSH and B. theta BSH). purity: 98%
Catalog No. 50-258-1702
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.