Learn More
Medchemexpress LLC BSJ-04-122 | 2513289-74-0 | 99.9% | 313.74 g/mol | C15H12ClN5O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15218510MG
Encompass_Preferred
BSJ-04-122 is a covalent dual inhibitor of the MAP kinase kinases MKK4 and MKK7 developed for research use. It demonstrates potent enzymatic inhibition (IC50 4 nM for MKK4; 181 nM for MKK7) and is applied in studies of cancer biology and MAPK signaling. The compound is supplied as a solid with high chemical purity and recommended storage conditions for both powder and solution forms.
Catalog No. 50-002-86406