Learn More
Market Lab Wire Basket For Flexycart 6 x 24.5x 14IN
Supplier:  Market Lab 2336
Encompass_Preferred
The Wire Basket For Flexycart MLI is a wire basket designed to fit on Flexycart MLI mobile instrument stands. It provides a convenient way to hold supplies and accessories. The basket measures 10"W x 15"L x 6"H and is made from chrome-plated steel wire with a black powder-coated steel frame. The basket attaches easily to the Flexycart stand with included hardware. Its open wire design allows users to see contents while providing ventilation. With a 150 lb weight capacity, this wire basket is a useful accessory for organizing equipment when using Flexycart MLI mobile stands.
Catalog No. NC2548591
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.