Learn More
ATCC Escherichia coli
Supplier:  ATCC 23228
Encompass
Escherichia coli strain Bs-8 is a whole-genome sequenced bacterium that was derived from an existing strain. This strain cannot repair UV lesions in DNA.
Catalog No. NC2302462
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.