Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BSND Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562853 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562853 Supplier Invitrogen™ Supplier No. PA562853
  • $654.00
    $541.65 / Each of 1

    Save $112.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: SPQQEPQGCR CPLDRFQDFA LIDAPTLEDE PQEGQQWEIA LPNNWQRYPR TKVEEKEASD TGGEEPEKEE EDLYYGLPDG AGDL Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 54%.

BSND encodes an essential beta subunit for CLC chloride channels. These heteromeric channels localize to basolateral membranes of renal tubules and of potassium-secreting epithelia of the inner ear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BSND
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BSND
Gene Accession No. Q8WZ55
Gene Alias BART; Bartter syndrome, infantile, with sensorineural deafness (Barttin); barttin; barttin CLCNK type accessory beta subunit; barttin CLCNK-type chloride channel accessory beta subunit; Bsnd; deafness, autosomal recessive 73; DFNB73
Gene Symbols BSND
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BSND. Recombinant protein control fragment (Product #RP-98526).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.