Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BSSP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114110 Supplier Invitrogen™ Supplier No. PA5114110
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPVPPACGKP QQLNRVVGGE DSTDSEWPWI VSIQKNGTHH CAGSLLTSRW For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 91%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 93%; Rat: 100%.

This gene encodes a member of the trypsin family of serine proteases. The enzyme is expressed in the airways in a developmentally regulated manner. The gene is part of a cluster of serine protease genes on chromosome 16.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BSSP4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRSS22
Gene Accession No. Q9GZN4
Gene Alias 4733401N09Rik; brain-specific serine protease 4; BSP-2; BSSP4; BSSP-4; hBSSP-4; MGC9599; prosemin; protease, serine 22; protease, serine S1 family member 22; protease, serine, 22; PRSS22; Prss26; Serine protease 22; serine protease 26; SP001LA; Tryptase epsilon; UNQ302/PRO343
Gene Symbols PRSS22
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human PRSS22 (aa 35-84).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64063
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.