Learn More
Invitrogen™ BST-1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total BST-1 / CD157 protein.
Specifications
Specifications
| Antigen | BST-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Bst1 |
| Gene Accession No. | Q10588, Q63072, Q64277 |
| Gene Alias | 114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase |
| Gene Symbols | Bst1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.