Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BSTFA Silylation Reagent

Catalog No. PI38828 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI38828 25 g
PI38829 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI38828 Supplier Thermo Scientific™ Supplier No. TS38828
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obtain excellent GC separations with Thermo Scientific™ BSTFA Silylation Reagent.

Improve GC analysis with Thermo Scientific™ BSTFA Silylation Reagent. The highly volatile byproducts mono(trimethylsilyl)trifluoroacetamide (BSTFA) and trifluoroacetamide elute with the solvent front to ensure excellent chromatographic separations. This silylation reagent reacts with a wide range of polar compounds and is particularly well suited for the GC analyses of some lower boiling TMS-amino acids and TMS Krebs cycle acids.

BSTFA is a powerful thrimethylsilyl donor, with donor strength that is comparable to its unfluorinated analog BSA [N,O-Bis(trimethylsilyl)acetamide]. BSTFA reacts to replace labile hydrogens on a wide range of polar compounds with a -Si(CH3)3 group. This physical characteristic is particularly useful in the gas chromatography of some lower boiling TMS-amino acids and TMS Krebs cycle acids.
  • Increased volatility of reaction byproducts mono(trimethylsilyl)trifluoroacetamide and trifluoroacetamide over corresponding nonfluorinated compounds from BSA
  • Increased volatility makes it possible to derivatize smaller molecules with which the TMS derivatives elute with the byproducts from BSA
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Storage 2°C to 8°C
Quantity 25 g
Standard Components Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate
Standard Concentration 98 to 100%

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.