Learn More
G-Biosciences Bsu DNA Polymerase, Large Fragment
SCI_ED
Catalog No. S03117
Change view
Click to view available options
No. of Reactions:
2000
Catalog No. No. of Reactions
S03117 2000
1 options
Catalog No. S03117 Supplier G-Biosciences Supplier No. 7862000
Add to Cart
Add to Cart
Bsu DNA Polymerase, Large fragment is used for second strand cDNA synthesis, strand displacement DNA synthesis, random primer labeling and dA-tailing.
  • Bsu DNA Polymerase Large fragment is from Bacillus subtilis and is modified to retain its 5' - 3' DNA polymerase activity with its 5' - 3' exonuclease domain removed.
  • It also lacks the 3' - 5' exonuclease activity.
Specifications
Reaction Speed Standard
Product Type DNA Polymerase
No. of Reactions 2000