Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Bsu DNA Polymerase, Large Fragment
SCI_ED

Catalog No. S03119
Change view
Click to view available options
No. of Reactions:
200
1000
2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03119 200
S03117 2000
S03118 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S03119 Supplier G-Biosciences Supplier No. 7861998
Add to Cart
Add to Cart

Bsu DNA Polymerase, Large fragment is used for second strand cDNA synthesis, strand displacement DNA synthesis, random primer labeling and dA-tailing.

  • Bsu DNA Polymerase Large fragment is from Bacillus subtilis and is modified to retain its 5' - 3' DNA polymerase activity with its 5' - 3' exonuclease domain removed.
  • It also lacks the 3' - 5' exonuclease activity.

Specifications

Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance High
PCR Method Standard PCR, qPCR, Multiplex qPCR, RT-PCR, 1-step RT-PCR
Reaction Speed Standard
Label or Dye Biotin, FAM, 6-FAM, SYBR Green, Unlabeled
Product Type DNA Polymerase
For Use With (Application) Microbial Detection, Pathogen Detection, Virus Detection, Genetic Disease Research, Isothermal Amplification
No. of Reactions 200
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.