Learn More
Bsu DNA Polymerase Large Fragment, Glycerol Free, G-Biosciences

Description
It is modified to retain its 5'-to-3' DNA polymerase activity with its 5'-to-3' exonuclease domain removed. It also lacks the 3'-to-5' exonuclease activity.
Specifications
Specifications
| Detection Method | Gel Electrophoresis |
| Form | Liquid |
| GC-Rich PCR Performance | High |
| PCR Method | End-point PCR |
| Reaction Speed | 20 min. |
| Label or Dye | Biotin, FAM, TAMRA |
| Product Type | DNA Polymerase |
| Quantity | 1000 U |
| For Use With (Application) | Microbial Detection, Pathogen Detection, Isothermal Amplification |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.