Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bsu DNA Polymerase Large Fragment, Lyophilized, G-Biosciences
SCI_ED

Catalog No. S000265
Change view
Click to view available options
Quantity:
1000 U
200 U
2000 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000265 1000 U
S000262 2000 U
S000275 200 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S000265 Supplier G-Biosciences Supplier No. 7862152
Add to Cart
Add to Cart

Bsu DNA Polymerase, large fragment is from Bacillus subtilis.

It is modified to retain its 5'-to-3' DNA polymerase activity with its 5'-to-3' exonuclease domain removed. It also lacks the 3'-to-5' exonuclease activity.

Specifications

Detection Method Gel Electrophoresis
Form Lyophilized
GC-Rich PCR Performance Medium
PCR Method End-point PCR
Reaction Speed 20 min.
Label or Dye Biotin, FAM, TAMRA
Product Type DNA Polymerase
Quantity 1000 U
For Use With (Application) Microbial Detection, Pathogen Detection, Isothermal Amplification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.