Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix Scientific BT2 Dip Slide
SCI_ED

Catalog No. S99496 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
10 Dip Slides/box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S99496 10 Dip Slides/box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S99496 Supplier AquaPhoenix Scientific Supplier No. BT2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nutrient Agar with TTC.

  • Nutrient Agar is a general purpose agar that tests for a total count of nonfastidious microorganisms.
  • The Nutrient agar incorporates red spot dye which makes it easier to interpret results.
  • The BT2 dipslide (TTC dipslide) is an all around general purpose dip slide that shows total count bacteria on both sides of the slide.

Specifications

Content And Storage 2°C to 8°C
Additive Triphenyl Tetrazolium Chloride Dye (TTC)
Media Type Nutrient Agar with TTC
Product Type Dip Slide
Target Organism Class Bacteria
Test Type Microbial Detection
Formulation Nutrient Agar with TTC (Red Spot Dye)
Incubation Conditions 30°C to 35°C
Incubation Time 24 to 48 hr.
No. per Pack 10/Pk.
Packaging Type Box
Quantity 10 Dip Slides/box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.