Learn More
TARGETMOL CHEMICALS INC BT44 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T677335MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BT44 is a novel second-generation glial cell line?derived neurotropic factor mimetic with improved biological activity and is a lead compound for the treatment of neurodegenerative disorders. purity: 100%
Catalog No. 50-257-9273
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.