Learn More
Abcam BTBCT, Europium Beta-Diketone Chelate, Solid, 5 mg
Description
- Molecular Weight: 604.9 Da
- Purity: >95%
- Description: Europium β-diketone chelate
- Performance: Provides 8 times improved detection sensitivity compared to BHHCT-Eu3+
- Form: Solid
- CAS Number: 525560-81-0
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% 4,4''-Bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4'-sulfonyl chloride |
| Content And Storage | Store at -20°C. |
| Quantity | 5 mg |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.