Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam BTBCT Europium Beta-Diketone Chelate, Solid, 604.9 Da, 250 mg
Description
- Molecular weight: 604.9 Da
- Purity: >95%
- Form: Solid
- Composition: 100% 4,4-Bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4'-sulfonyl chloride
- Key benefits: Provides 8 times improved detection sensitivity compared to BHHCT-Eu³+
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Label or Dye | Europium Chelate |
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Buffer | 100% 4,4''-bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4'-sulfonyl chloride |
| Formulation | 100% 4,4''-Bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4'-sulfonyl chloride |
| Quantity | 250 mg |
| For Use With (Application) | Sample Prep and Detection |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?