Learn More
Abcam BTBCT, Europium Beta Diketone Chelate, Solid, >95% Purity, 50 mg
Description
- Molecular Weight: 604.9 Da
- Purity: >95%
- Form: Solid
- Chemical Class: Europium β-diketone chelate
- Composition: 100% 4,4-Bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4'-sulfonyl chloride
- Performance: Provides 8 times improved detection sensitivity compared to BHHCT-Eu³+
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% 4,4''-Bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4'-sulfonyl chloride |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Label or Dye | Europium Chelate |
| Detection Method | Fluorescence |
| Formulation | 100% 4,4''-Bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4'-sulfonyl chloride |
| Quantity | 50 mg |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.