Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTEX Solution, MilliporeSigma™ Supelco™

Catalog No. 111002692 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2692 10 x 1 mL
11-100-2693 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-2692 Supplier MilliporeSigma Supelco Supplier No. 4372810X1ML
Add to Cart
Add to Cart

BTEX are important chemicals, which are commonly used as chemical intermediates and solvents in the production of plastics.{11)

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-88-3
Density 1.099 g/mL (at 20°C)
Type Analytical Standard
Quantity 10 x 1 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.