Learn More
Invitrogen™ BTG1 Monoclonal Antibody (OTI7H6)

Description
BTG1 Monoclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | BTG1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI7H6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | BTG1 |
| Gene Accession No. | P62324 |
| Gene Alias | AI426953; anti-proliferative factor; APRO2; AW546738; B cell translocation gene 1, anti-proliferative; B-cell translocation gene 1 protein; B-cell translocation gene 1, anti-proliferative; BTG anti-proliferation factor 1; Btg1; Protein BTG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.