Learn More
Invitrogen™ BTG3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total BTG3 protein.
Specifications
Specifications
| Antigen | BTG3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | BTG3 |
| Gene Accession No. | P50615, Q14201 |
| Gene Alias | Abundant in neuroepithelium area protein; ANA; APRO4; B cell translocation gene 3; B-cell translocation gene 3; BTG anti-proliferation factor 3; BTG family member 3; BTG family, member 3; Btg3; btg3 {ECO:0000312; Protein BTG3; protein Tob5; rBTG3; RGD:2226}; TOB5; TOB55; TOFA |
| Gene Symbols | BTG3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.