Learn More
Market Lab Bottle Thermometers, ML20101, 3ml
Supplier:  Market Lab ML20102
Encompass_Preferred
The MLI ML20101 Bottle Thermometer is a glass thermometer with a blue alcohol-filled column and white numbers for easy reading. It has a temperature range of -20 to 110°F/-28 to 43°C and is 8 inches long. This thermometer is designed for use in liquids and attaches to bottles, carboys, and tanks with a stainless steel clip. It can be fully submerged and is ideal for monitoring fermentation or controlling bath temperatures. The thermometer has 1° graduations for precise temperature measurement. It is made in the USA and is lead-free and non-toxic.
Catalog No. NC2739054
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.