Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix Scientific BTM2 Dip Slide
SCI_ED

Catalog No. S99497 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Additive:
Triphenyl Tetrazolium Chloride Dye (TTC)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Additive
S99497 Triphenyl Tetrazolium Chloride Dye (TTC)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S99497 Supplier AquaPhoenix Scientific Supplier No. BTM2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nutrient Agar with TTC and Malt Extract Agar.

  • Nutrient Agar is a general purpose agar that tests for a total bacteria count.
  • The Nutrient agar incorporates red spot dye which makes it easier to interpret results.
  • Malt Extract Agar tests for yeasts and molds.
  • This is a general purpose slide that can be used in a variety of applications.

Specifications

Content And Storage 2°C to 8°C
Additive Triphenyl Tetrazolium Chloride Dye (TTC)
Detects Total bacteria count
Media Type Nutrient Agar with TTC and Malt Extract Agar
Product Type Dip Slide
Test Type Microbial Detection
Formulation Nutrient Agar with TTC (Red Spot Dye), Malt Extract Agar
For Use With (Application) General Hygiene Monitoring
Incubation Conditions 30°C
Incubation Time 24 to 120 hr.
No. per Pack 10/Pk.
Packaging Type Box
Specimen Type Food, Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.