Learn More
OriGene BTN3A1 Human shRNA Plasmid Kit (Locus ID 11119)
SDP
Supplier:  OriGene TL316960
Encompass_Preferred
BTN3A1 - Human, 4 unique 29mer shRNA constructs in lentiviral GFP vector, 5µg of each construct provided
Catalog No. NC2743709