Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTNL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7210938 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4728 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4728 Supplier Proteintech Group Inc Supplier No. 251101AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTNL2 (butyrophilin-like protein 2), also known as BTL-II, is a 455 amino acid single-pass type II membrane protein that acts as a negative regulator of T cell proliferation. Expressed in thymus, brain, kidney, heart, liver, small intestine, testis, ovary, pancreas and leukocytes, BTNL2 exists as at least six alternatively spliced isoforms, which are encoded by a gene located on human chromosome 6. Expression of BTNL2 isoform 3 is associated with susceptibility to sarcoidosis type 2 (SS2), an immune disorder that causes chronic inflammatory granulomatous lesions.

Specifications

Antigen BTNL2
Applications Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BTNL2
Gene Accession No. O70355, Q9UIR0
Gene Alias BTL II, BTNL2, Butyrophilin like protein 2, HSBLMHC1, SS2
Gene Symbols Btnl2
Host Species Rabbit
Immunogen BTNL2 Fusion Protein Ag18803
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 547431, 56244
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.