Learn More
Perkin Elmer US LLC BTO Orange Injection Port Septa, Pkg. 10
Supplier:  Perkin Elmer US LLC N9306872
Encompass_Preferred
Septa rated to 400 °C. The precision molded silicone rubber septa BTO (Bleed Temperature Optimized) are premium ultra-low bleed injector septa for todays most demanding applications.
Catalog No. 50-211-2757