Learn More
TARGETMOL CHEMICALS INC 740 Y-P 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ00035MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. 740 Y-P (740YPDGFR) is a potent and cell-permeable activator of PI3K. purity: 99%
Catalog No. 50-257-8849
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.