Learn More
Selleck Chemical LLC BTYNB-25mg
SDP
Supplier:  Selleck Chemical LLC S987125MG
Encompass_Preferred
BTYNB (BTYNB IMP1 Inhibitor, MDK6620) is a potent and selective inhibitor of IMP1 binding to c-Myc mRNA. BTYNB downregulates β-TrCP1 mRNA and reduces activation of nuclear transcriptional factors-kappa B (NF-κB). BTYNB disrupts this enhancer function by impairing IGF2 mRNA-binding protein 1 (IGF2BP1)-RNA association.
Catalog No. 50-238-9657
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.