Learn More
Abcam Bu 224 Hydrochloride, Imidazoline I2 Binding Site Modulator, Solid, 10 mg
Description
- Molecular Weight: 233.69 Da
- Purity: >99%
- Form: Solid
- Activity: High affinity Imidazoline I2 binding site allosteric modulator (Ki = 2.1 nM)
- Functional Effects: Dose-dependently inhibits C-fiber evoked responses, post-discharge and wind-up; shows antinociceptive effects in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Imidazoline I2 Binding Site |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% BU 224 hydrochloride |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Quantity | 10 mg |
| For Use With (Application) | Neuroscience-Antinociception research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.