Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam BU 224 Hydrochloride, Imidazoline I2 Binding Site Modulator, Solid, 50 mg

Catalog No. 502704636
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4636 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4636 Supplier Abcam Supplier No. AB14641550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-affinity allosteric modulator of imidazoline I2 binding sites with antinociceptive effects.

  • Molecular Weight: 233.69 Da
  • Purity: >99%
  • Form: Solid
  • Mechanism: High affinity Imidazoline I2 binding site allosteric modulator (Ki = 2.1 nM)
  • Biological Activity: Dose-dependently inhibits C-fiber evoked responses, post-discharge and wind-up; shows antinociceptive effects in vivo
  • Composition: 100% BU 224 hydrochloride

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Imidazoline I2 Binding Site
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Format Vial
Buffer 100% BU 224 hydrochloride
Content And Storage Store at ambient temperature.
Form Solid
Formulation 100% BU 224 hydrochloride
Quantity 50 mg
For Use With (Application) Neuroscience-Antinociception research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.