Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUB1 Mouse anti-Human, Clone:1A11C7, Abnova™

Catalog No. 89334495 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-495 Supplier Abnova Corporation Supplier No. MAB17852
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human BUB1.

This gene encodes a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint. Mutations in this gene have been associated with aneuploidy and several forms of cancer. [provided by RefSeq]

Specifications

Antigen BUB1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1A11C7
Conjugate Unconjugated
Description budding uninhibited by benzimidazoles 1 homolog (yeast)
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene BUB1
Gene Alias BUB1A, BUB1L, hBUB1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-130 of human BUB1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 699
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.