Learn More
Synthware™ Bubbler
GREENER_CHOICESDP
Catalog No. 31500703 Shop All Kemtech Products
Change view
Click to view available options
Includes:
Ground Joint on the Bottom
Vacuum Port on the Bottom
Standard Taper Joint:
29/42
29/42, 14/20
29/42, 24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Standard Taper Joint
31-500-703 Ground Joint on the Bottom 29/42, 14/20
31-500-704 Ground Joint on the Bottom 29/42, 24/40
31-500-705 Vacuum Port on the Bottom 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-703 Supplier Synthware™ Supplier No. B262914
Add to Cart
Add to Cart
Bubbler has a small ball joint to prevent oil from being sucked back under sudden negative pressure
Specifications
Material Borosilicate Glass
Type Mineral Oil Bubbler
Includes Ground Joint on the Bottom
Standard Taper Joint 29/42, 14/20
Length (Metric) 157 mm
For Use With (Application) Organic Synthesis
Quantity 1 Each