Learn More
BUCHI Boiling Point Tubes and Capillaries
SDP
Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.
$117.95 - $1093.27
Specifications
Material Borosilicate Glass 3.3
Autoclavable Autoclavable
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Packaging Quantity Price Quantity & Availability  
05-402-793
SDP
View Documents
BUCHI
019007
Boiling Point Tubes 100/Pk.
Each for $1,093.27
Only null left
Add to Cart
05-402-814
SDP
View Documents
BUCHI
019697
Boiling Point Tubes 10/Pk.
Each for $117.95
Only null left
Add to Cart
10-987-108
SDP
View Documents
BUCHI
051850
Boiling Point Capillaries 10/Pk.
Pack of 10 for $387.79
Only null left
Add to Cart
Description
Description
  • High precision for reliable results
  • Meets Siwoloboff and Eu Pharmacopeia standards
  • Designed for BUCHI melting and boiling point instruments
  • Compatible with other instruments
Specifications
Specifications
Borosilicate Glass 3.3
Autoclavable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.