Learn More
BUCHI FlashPure EcoFlex Silica TLC Plates
SDP
Catalog No. 05405559 Shop All BUCHI Products
Change view
Click to view available options
Optional Vacuum Controller V-850
Catalog No.
05-405-559 Optional Vacuum Controller V-850
1 options
Catalog No. 05-405-559 Supplier BUCHI Supplier No. 140000174
Glass or aluminum backed plates coated with FlashPure Ecoflex Silica with fluorescent indicator, UV 254 nm.
Specifications
Backing Material Glass backed
Type TLC Plate, Glass backed (UV 254nm) 20 x 20cm