Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Budget Skeleton
SCI_ED

Catalog No. SCH10 Shop All Walter Products Products
Change view
Click to view available options
Budget skeleton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SCH10 Budget skeleton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCH10 Supplier Walter Products™ Supplier No. B10201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Life-size and affordable.

This economical, life-size articulated adult plastic skeleton is ideal for teaching the basics of anatomy.

  • Removable arms and legs
  • Includes nerve branches, vertebral artery and herniated lumbar disk
  • Movable jaw, cut calvarium, suture lines and three removable lower teeth
  • Complete with dust cover and skeletal system chart
  • Mounted on a sturdy, 16 in. (40cm) wide metal base
  • Height: 5 ft. 6 in. (1.68m)

Specifications

For Use With (Application) Ideal for teaching the basics of anatomy
Height (English) 66.14 in.
Height (Metric) 168 cm
Includes Nerve branches, vertebral artery and herniated lumbar disk, skull features movable jaw, suture lines, three movable teeth and calvarium, dust cover and skeletal system chart, removable arms and legs
Material Plastic
Mounting On a sturdy, 16 in. W Metal base
Product Type Skeleton Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.