Learn More
Selleck Chemical LLC Bufalin
SDP
Supplier:  Selleck Chemical LLC S78215MG
Encompass_Preferred
Bufalin (BF) is a potent inhibitor of steroid receptor coactivator SRC-3 SRC-1 and Na /K -ATPase Bufalin binds to the subunit 1 2 and 3 of Na /K -ATPase with Kd of 42 5 nM 45 nM and 40 nM respectively Bufalin is a major immunoreactive component isolated from the Chinese medicine Chan Su with anti-cancer activity
Catalog No. 50-217-3310
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.