Learn More
Thomas Scientific 10x PCR Buffer Mg free for Choice-TaqT DNA Polymerase, 1.0mL
SDP
Supplier:  Thomas Scientific C788T64
Encompass
Choice-Taq™ DNA Polymerase is a recombinant Taq DNA polymerase Choice-Taq™ is supplied with a 10x NH4 Reaction buffer containing 1.5mM MgCI2 (final concentration)
Catalog No. 50-145-9096
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.