Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Buffer Pad

Catalog No. 19072004 Shop All 3M Products
Change view
Click to view available options
Diameter (English):
13 in.
20 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
19-072-004 20 in.
19-072-011 13 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-004 Supplier 3M™ Supplier No. 7000000663
Add to Cart
Add to Cart

3M™ Red Buffer Pad quickly cleans and removes scuff marks and enhances floor appearance.

  • Use to light scrub without dulling the finish
  • Can be used for everyday cleaning under and automatic scrubber
  • Specifically designed for spray buffing
  • Cleans when damp; Buffs when dry
  • Use on rotary or automatic floor machines
  • Works well on automatic floor scrubbers for light duty cleaning
TRUSTED_SUSTAINABILITY

Specifications

Product Type Buffer Pad
Color Red
Diameter (English) 20 in.
Diameter (Metric) 510 mm
For Use With (Application) Spray Cleaning Protected Floors
Material Non-Woven Polyester Fiber
Model 5100-20
Quantity 5/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.