Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, pH 10.5, Reagents

Catalog No. 01335728
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-728 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-728 Supplier Reagents Holdings Llc Supplier No. CB205000500A
Add to Cart
Add to Cart

Buffer, pH 10.5, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
pH 10.5
Physical Form Liquid
Quantity 500 mL
Packaging Poly Bottle
Sterility Non-sterile
Solubility Information Completely miscible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.